Learn More
Apexbio Technology LLC Fenofibrate 49562-28-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19435.1
Encompass_Preferred
Fenofibrate is a selective agonist targeting the peroxisome proliferator-activated receptor-alpha (PPAR ) a nuclear receptor involved in lipid metabolism regulation Upon binding to PPAR Fenofibrate modulates gene transcription associated with fatty acid oxidation and lipid homeostasis In vitro assays indicate Fenofibrate activates mouse and human PPAR receptors at EC50 values of approximately 18 M and 30 M respectively Commonly employed in biomedical research Fenofibrate serves as a pharmacological tool in studies investigating lipid disorders metabolic syndrome cardiovascular diseases and related metabolic pathways under experimental conditions
Catalog No. 50-000-62492