Learn More
Medchemexpress LLC Fenofibrate | 49562-28-9 | 99.9% | 360.83 | 1 ML

Supplier: Medchemexpress LLC HY1735610MM/1ML
Fenofibrate is a selective peroxisome proliferator-activated receptor alpha (PPARα) agonist with an EC50 of 30 μM. It also demonstrates inhibitory activity against various human cytochrome P450 isoforms. This compound is widely utilized in cardiovascular disease and cancer research.
- Selective PPARα agonist
- Inhibits human cytochrome P450 isoforms (CYP2C19, CYP2B6, CYP2C9, CYP2C8, CYP3A4)
- Useful in cardiovascular disease and cancer research
- Supplied as a 10 mM solution in 1 mL DMSO
- Appearance is a solid with a melting point of 80-81°C
- High purity of 99.92%
- Stable for 3 years as powder at -20°C or 1 year in solvent at -80°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.