Learn More
Medchemexpress LLC Fenofibrate | 49562-28-9 | 99.9% | 360.83 | 10 G
SDP
Supplier:  Medchemexpress LLC HY1735610G
Encompass_Preferred
Fenofibrate is a selective PPARα agonist with an EC50 of 30 μM. It also inhibits human cytochrome P450 isoforms, specifically CYP2C19, CYP2B6, CYP2C9, CYP2C8, and CYP3A4, with IC50s of 0.2, 0.7, 9.7, 4.8, and 142.1 μM, respectively.
  • Selective PPARα agonist.
  • Inhibits multiple human cytochrome P450 isoforms.
  • Available as a reference standard, deuterium-labeled, and GMP grade.
Catalog No. 50-003-84472