Learn More
Medchemexpress LLC Fenofibrate (Standard) | 49562-28-9 | 99.9% | 360.83 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17356R50MG
Encompass_Preferred
Fenofibrate (Standard) is the analytical standard of Fenofibrate, intended for research and analytical applications. It is a selective PPARα agonist and inhibits human cytochrome P450 isoforms. This analytical standard is supplied for assay purposes and is commonly used in various research experiments.
  • For research and analytical applications
  • Selective PPARα agonist
  • Inhibits human cytochrome P450 isoforms
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-84625