Learn More
Apexbio Technology LLC Fenoldopam 67227-56-9 10mg
SDP
Supplier:  Apexbio Technology LLC B148310
Encompass_Preferred
Fenoldopam is a small-molecule partial agonist targeting dopamine D1-like receptors It is designed to selectively activate these receptors thereby mediating peripheral vasodilation and modulating vascular resistance Fenoldopam exerts its biological activity primarily through D1-like receptor activation In in vitro studies Fenoldopam demonstrates partial agonist activity with an EC50 value of approximately 57 nM Additionally it exhibits inhibitory activity against 2-adrenoceptors with a reported Ki of 15 25 nM Based on these pharmacological properties Fenoldopam holds research potential in studies of cardiovascular regulation receptor pharmacodynamics and selective receptor ligand profiling
Catalog No. 50-000-89965