Learn More
Medchemexpress LLC Fenoldopam mesylate | 67227-57-0 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0735A100MG
Encompass_Preferred
Fenoldopam mesylate functions as a D1 receptor agonist and a novel lysine-specific demethylase 1 (LSD1) inhibitor with an IC50 of 0.8974 μM. This compound demonstrates anti-hypertensive effects, exhibits anti-cancer cell proliferation activity, and can induce cell apoptosis, making it a subject of research in various biological contexts.
  • Functions as a D1 receptor agonist
  • Acts as a novel LSD1 inhibitor
  • Exhibits anti-hypertensive properties
  • Shows anti-cancer cell proliferation activity
  • Capable of inducing cell apoptosis
  • Active against RCC cell lines (786-O, ACHN) in vitro
Catalog No. 50-004-05260