Learn More
Selleck Chemical LLC Fenoldopam mesylate S4618-10mg
SDP
Supplier:  Selleck Chemical LLC S461810MG
Encompass_Preferred
Fenoldopam (SKF 82526 Corlopam) is a selective dopamine-1 receptor (DA1) agonist with natriuretic/diuretic properties It lowers blood pressure through arteriolar vasodilation
Catalog No. 50-310-8907