Learn More
Cayman Chemical Fenoldopamesylate 10mg
SDP
Supplier:  Cayman Chemical 1762910 MG
Encompass_Preferred
An agonist of dopamine D1A (D1R) and D1B (D5R) receptors (Kds = 17 and 11 nM, respectively); alters hemodynamic properties, including hypertension, in animals.
Catalog No. 50-296-5851