Learn More
Medchemexpress LLC Fenoverine | 37561-27-6 | MFCD00865242 | 99.7% | 459.56 g·mol⁻¹ | C26H25N3O3S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY107349100MG
Encompass_Preferred
Fenoverine is an antispasmodic research compound (CAS 37561-27-6) that modulates smooth muscle activity by inhibiting voltage-dependent calcium channels. It is provided as a high-purity analytical standard for preclinical pharmacology and analytical applications.
  • Antispasmodic compound for smooth muscle research.
  • Reduces calcium ion influx via voltage-dependent channel inhibition.
  • High purity: 99.7%.
  • Molecular formula C26H25N3O3S; molecular weight 459.56 g·mol⁻¹.
  • Available as solid powder and as a DMSO solution for biochemical assays.
Catalog No. 50-002-40875