Learn More
Medchemexpress LLC Fenpropidin | 67306-00-7 | 99.5% | 273.46 g/mol | C19H31N | 5MG

Supplier: Medchemexpress LLC HY1262005MG
Fenpropidin is a piperidine-class fungicide provided as a research reagent for in vitro and in vivo studies; it exhibits antifungal activity against various yeasts and filamentous fungi and includes recommended solubility and storage information for experimental use.
- High purity (99.5%).
- Chemical formula C19H31N; molecular weight 273.46 g/mol.
- CAS number 67306-00-7.
- Soluble in DMSO (100 mg/mL); several in vivo vehicle formulations reach ≥2.5 mg/mL.
- Storage: protect from light; store at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
- Available in small-scale quantities and solution formats (for example, 1 mg, 5 mg, 10 mg, and 10 mM in DMSO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.