Learn More
Medchemexpress LLC Fenpropimorph | 67564-91-4 | MFCD00144306 | 98.7% | 303.48 g/mol | C20H33NO | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1280335MG
Encompass_Preferred
Fenpropimorph is an agricultural fungicide provided for research and analytical use. It inhibits the sterol biosynthesis pathway and is supplied as a colorless to light-yellow liquid with high purity suitable for reference and experimental work. Typical specifications include CAS 67564-91-4, molecular formula C20H33NO, molecular weight 303.48 g/mol, and purity ≈98.74%.
  • Inhibits sterol biosynthesis, useful for studies of membrane sterols.
  • High purity (≈98.7%) suitable for analytical applications.
  • Available in small research quantities for laboratory use.
  • Soluble in DMF at approximately 15 mg/mL with warming and sonication.
  • Colorless to light-yellow liquid with documented density.
  • Suitable as a reference standard in biochemical and toxicological assays.
Catalog No. 50-002-64661