Learn More
Medchemexpress LLC Fenretinide | 65646-68-6 | MFCD00792674 | C26H33NO2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15373
Encompass_Preferred
Fenretinide (4-HPR) is a synthetic retinoid derivative that binds to retinoic acid receptors (RAR) at concentrations necessary to induce cell death. It is intended for research use only.
  • Exerts acute and long-term antitumor activity in selected T-ALL cell lines.
  • Inhibits DES activity in CCRF-CEM leukemia cells, leading to increased dhCer content.
  • Protects insulin signaling by preventing lipid-induced reductions in insulin-stimulated glucose uptake.
  • Inhibits OVCAR-5 cell proliferation and viability.
  • Prevents endothelial cells from forming tubes at 1 microM.
  • Selectively inhibits ceramide accumulation in HFD-fed male C57Bl/6 mice.
  • Improves glucose tolerance and insulin sensitivity in mice.
  • Ketoconazole can increase 4-HPR plasma levels in NOD/SCID mice.
Catalog No. 50-003-75607