Learn More
Apexbio Technology LLC Fenspiride HCl 5053-08-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B17545.1
Encompass_Preferred
Fenspiride hydrochloride (CAS 5053-08-7) is a small molecule antagonist of -adrenergic and histamine H1 receptors By inhibiting these signaling pathways Fenspiride HCl modulates bronchial smooth muscle tone and reduces inflammatory processes mediated by histamine and adrenergic activity This compound is utilized in biomedical research to investigate mechanisms of airway hyperreactivity neurogenic inflammation and the pharmacological modulation of respiratory tract disorders related to histaminergic and adrenergic signaling
Catalog No. 50-000-94665