Learn More
Apexbio Technology LLC Fenticonazole Nitrate 73151-29-8 50mg
SDP
Supplier:  Apexbio Technology LLC A843250
Encompass_Preferred
Fenticonazole Nitrate (CAS 73151-29-8) is a small-molecule inhibitor targeting fungal cytochrome P450 enzyme CYP51A1 (lanosterol 14 -demethylase) It is designed to inhibit the demethylation step required for ergosterol biosynthesis thereby disrupting fungal cell membrane integrity and growth Fenticonazole Nitrate exerts its biological activity primarily through inhibition of CYP51A1 In experimental studies Fenticonazole Nitrate demonstrates inhibitory activity with reported IC50 values for CYP51 inhibition typically in the low micromolar concentration in fungal models such as Malassezia furfur and Candida albicans Based on these pharmacological properties Fenticonazole Nitrate holds research potential in antifungal modes of action studies and antifungal screening assays
Catalog No. 50-000-94487