Learn More
Medchemexpress LLC Fenticonazole Nitrate | 73151-29-8 | 98.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB035910MM/1ML
Encompass_Preferred
Fenticonazole Nitrate is an antifungal imidazole ring derivative. It functions by hindering ergosterol integration, which in turn destructs the cytoplasmatic outer membrane. Fenticonazole Nitrate is effective against Gram-positive bacteria, mycoses, and vaginal candidiasis.
  • Antifungal imidazole ring derivative
  • Hinders ergosterol integration
  • Destroys the cytoplasmatic outer membrane
  • Effective against Gram-positive bacteria
  • Effective against mycoses
  • Effective against vaginal candidiasis
Catalog No. 50-004-05312