Learn More
Accuflow Systems Saint-Gobain Versilon ConvoFlex Tubing, FEP, Convoluted, 8 ft
Supplier:  Accuflow Systems FXFE669003096
Encompass_Preferred
Versilon ConvoFlex convoluted FEP tubing designed for flexible, chemically resistant fluid transfer and conduit applications. The helically convoluted profile provides excellent flexibility and a spiral design that aids self-flushing and cleaning, while FEP construction delivers chemical resistance, elevated temperature performance, and read-through transparency. Supplied in an 8 ft length.
Catalog No. NC2659832