Learn More
Chemglass Life Sciences FEP Tubing; 1" OD x 7/8" ID x 10ft Long (Supplied as two 5ft lengths)
SDP
Supplier:  Chemglass Life Sciences CG197514
Encompass_Preferred
FEP Tubing; 1" OD x 7/8" ID x 10ft Long (Supplied as two 5ft lengths). FEP heavy wall tubing for use with Chemglass process reactors.
Catalog No. NC1478327