Learn More
Bellco Glass, Inc. FermProbeモ pH Probe,12x325mm Broadley-James
SDP
Supplier:  Bellco Glass, Inc. 780322520
Encompass_Preferred
The Fermprobe pH electrode with S8 metric cap features a standard S8 detachable cable connector on a metric threaded polymer cap. The design requires the rotation of the electrode during installation this style of pH. Fermprobe is the first choice for small autoclavable bench-top vessels. All Fermprobe pH electrodes are designed to be used in bio-process applications where clean in place sterilize in place CIP, SIP procedures are used. Built to withstand repeated steamsterilization cycles the Fermprobe quickly stabilizes after steam exposure to be on line in half the time of other electrodes. This rugged low-impedance pH electrode continues to provide fast and precise measurement.--Metric Pg13.5 threaded disconnect cap--pH Range: 0..13--Steam sterilizable to 135°C--150 psig max. pressure--Double junction, Ag-AgCl reference system
Catalog No. 50-194-6300