Learn More
Sino Biological Ferret CD4 Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK6000315
Encompass_Preferred
The Ferret CD4 ELISA Pair Set is for the quantitative determination of Ferret CD4.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Ferret CD4 coated on a 96-well plate. Standards and samples are added to the wells, and any Ferret CD4 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Ferret CD4 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Ferret CD4 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85997