Learn More
Medchemexpress LLC Ferric carboxymaltose | 53858-86-9 | 10 MG

Supplier: Medchemexpress LLC HY108765100MG
Ferric carboxymaltose is an iron supplement intended exclusively for research purposes. It is not approved or sold for patient use.
- Supplied as a solid.
- Appearance ranges from brown to reddish brown.
- Store at 4°C, sealed and away from moisture.
- Suitable for biochemical assays.
- Used in research related to blood or cardio-cerebrovascular disease.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.