Learn More
Medchemexpress LLC Ferric ferrocyanide | 14038-43-8 | MFCD00135663 | ≥99.0% | 859.23 g/mol | C18Fe7N18 | 50 G
SDP
Supplier:  Medchemexpress LLC HY106594A50G
Encompass_Preferred
Prussian blue insoluble is an inorganic ferric ferrocyanide powder used as an adsorbent and research reagent in toxicology, imaging, and materials applications. It is commonly used to sequester cesium and thallium ions, serve as a pigment, and act as a contrast agent in select imaging studies.
  • Insoluble ferric ferrocyanide powder for laboratory use.
  • High purity (≥99.0% by assay).
  • Molecular weight 859.23 g/mol.
  • Effective adsorbent for cesium and thallium ions.
  • Applicable as a contrast agent in photoacoustic and MRI research.
  • Supplied as a dry powder suitable for experimental preparation.
Catalog No. 50-002-40509