Learn More
Medchemexpress LLC Ferric maltol | 33725-54-1 | MFCD33402141 | 99.9% | 431.15 | C18H15FeO9 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10801710MM 1ML
Encompass_Preferred
Ferric maltol is an orally active ferric (Fe3+) complex used in research on iron deficiency anemia. It increases transferrin saturation and serum iron-related biomarkers, while chelation reduces the presence of free iron in the gut, potentially lowering local toxicity compared with unchelated iron preparations.
  • Orally active ferric (Fe3+) complex for anemia research.
  • Increases transferrin saturation and serum iron markers.
  • Chelated iron minimizes free iron-mediated intestinal toxicity.
  • Available as solid or pre-dissolved solution for convenience.
  • High purity suitable for laboratory studies.
Catalog No. 50-002-41741