Learn More
Medchemexpress LLC HY-108017 5mg Medchemexpress, Ferric maltol CAS:33725-54-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1080175MG
Encompass_Preferred
Medchemexpress, HY-108017 5mg Ferric maltol CAS:33725-54-1 Ferric maltol is an orally active complex of a single ferric ion (Fe3+). Ferric maltol has tha potential for iron deficiency anemia treatment in inflammatory bowel disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0728