Learn More
Medchemexpress LLC Ferrichrome Iron-free (Desferrichrome) | 34787-28-5 | MFCD28899080 | 99.0% | 687.70 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP19975MG
Encompass_Preferred
Ferrichrome is a hydroxamate siderophore produced by various fungi (e.g., U. sphaerogena) that facilitates iron chelation and uptake. It can also function as a heterosiderophore for bacteria like Pseudomonas aeruginosa and Vibrio parahaemolyticus. Ferrichrome (0.8 μM) has been shown to inhibit concanavalin A-induced proliferation of mouse spleen monocytes and reduce the number of concanavalin A-stimulated CD4+ T cells expressing the IL-2 receptor. Additionally, it inhibits the heme-catalyzed oxidation of LDL by hydrogen peroxide in a concentration-dependent manner.
  • Facilitates iron chelation and uptake by various fungi.
  • Can be used as a heterosiderophore by bacteria.
  • Inhibits concanavalin A-induced proliferation of mouse spleen monocytes.
  • Reduces concanavalin A-stimulated CD4+ T cells expressing IL-2 receptor.
  • Inhibits heme-catalyzed oxidation of LDL by hydrogen peroxide.
Catalog No. 50-004-22921