Learn More
Cayman Chemical FerrIstatIn II 100g
SDP
Supplier:  Cayman Chemical 36621100 G
Encompass_Preferred
An iron uptake inhibitor; inhibits iron uptake (IC50 = ~12 µM), as well as induces degradation of TfR1 in HeLa cells; reduces serum iron levels and transferrin saturation in rats at 0.2, 10, and 40 mg/kg; suppresses traumatic brain injury-induced lipid peroxidation, neuronal apoptosis, and the number of cortical iron deposits in rats
Catalog No. 50-291-0076