Learn More
Medchemexpress LLC Ferrocene, 1,1'-bis(diphenylphosphino)- | 12150-46-8 | MFCD00001422 | 98.0% | 554.38 g·mol⁻¹ | C34H28FeP2 | 500g
SDP
Supplier:  Medchemexpress LLC HYY1237500G
Encompass_Preferred
DPPF (1 1 -Ferrocenediyl-bis(diphenylphosphine)) is a metal ligand DPPF enhances the stability of metal complexes DPPF can be used in the research of ovarian cancer[1][2][3]
Catalog No. 50-002-66976