Learn More
Medchemexpress LLC Ferrocene-d10 | 12082-87-0 | 99.1% | 196.09 g/mol | C10D10Fe | 10 MG
SDP
Supplier:  Medchemexpress LLC HY79504S10MG
Encompass_Preferred
Ferrocene-d10 is a deuterium-labeled ferrocene used as an isotope-labeled standard in analytical and research workflows. It provides a stable, high-purity reference for quantitative analyses and spectroscopic experiments.
  • Deuterium-labeled ferrocene with formula C10D10Fe.
  • High purity suitable for analytical use: 99.1%.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Solid, light yellow to brown appearance; handle and store protected from light under inert atmosphere.
  • Available in small pack sizes for trace-level and method development work.
Catalog No. 50-002-98819