Learn More
Cayman Chemical FerroLOXIN-1 10mg
SDP
Supplier:  Cayman Chemical 3932510 MG
Encompass_Preferred
A 15-LO-2/PEBP1 and ferroptosis inhibitor; selectively inhibits the peroxidation of 1-stearoyl-2-arachidonoyl-sn-glycero-3-PE by the 15-LO-2/PEBP1 complex over 15-LO-2 alone in cell-free assays at 0.25 µM; inhibits RSL3-induced ferroptosis in primary human bronchial epithelial cells (IC50 = 84 nM); increases survival in a mouse model of total body irradiation at 25 mg/kg
Catalog No. 50-291-7872