Learn More
Cayman Chemical FerroprotoporphyrIn 50mg
SDP
Supplier:  Cayman Chemical 3432750 MG
Encompass_Preferred
An iron-containing cofactor; involved the transport or storage of oxygen by hemoglobin or myoglobin, respectively, electron transfer by cytochrome b5, and oxidation of xenobiotics or endogenous substrates by CYPs
Catalog No. 50-295-0707