Learn More
Cayman Chemical FerroptosIs InhIbItr D1 5mg
SDP
Supplier:  Cayman Chemical 418225 MG
Encompass_Preferred
A free radical scavenger and an inhibitor of ferroptosis; scavenges 41.27% of DPPH radicals in a cell-free assay at 0.4 µM; inhibits erastin-induced lipid peroxidation in HT-1080 cells at 0.5 µM and erastin-induced ferroptosis in the same cells (IC50 = 22 nM); reduces infarct volume and improves neurological deficits in a mouse model of transient MCAO-induced ischemic brain injury at 5 mg/kg
Catalog No. 50-294-2617