Learn More
TARGETMOL CHEMICALS INC FERROSTATIN-1 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T650025MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ferrostatin-1 (Fer-1) is a potent ferroptosis inhibitor (EC50 60 nM). purity: 99%
Catalog No. 50-257-9346