Learn More
Medchemexpress LLC HY-100579 50mg Medchemexpress, Ferrostatin-1 CAS:347174-05-4 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY10057950MG
Encompass_Preferred
Medchemexpress,HY-100579 50mg Ferrostatin-1 CAS:347174-05-4 Formula:C15H22N2O2 Ferrostatin-1 is a potent inhibitor of ferroptosis with an EC50 of 60 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-6781