Learn More
Selleck Chemical LLC Ferrostatin-1 (Fer-1) 5mg 347174-05-4
SDP
Supplier:  Selleck Chemical LLC S72435MG
Encompass_Preferred
Ferrostatin-1 (Fer-1) is a potent and selective inhibitor of ferroptosis with EC50 of 60 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4553