Learn More
Medchemexpress LLC Ferrous bisglycinate | 20150-34-9 | 98.0% | 50 G
SDP
Supplier:  Medchemexpress LLC HY13007850G
Encompass_Preferred
Ferrous bisglycinate is an orally active iron fortificant and a therapeutic iron supplement. It is utilized in the research of iron deficiency anemia.
  • Orally active
  • Iron fortificant
  • Therapeutic iron supplement
  • Primarily used for the research of iron deficiency anemia
  • Did not affect Caco-2 cell viability at 25-200 μM for 2 hours
  • Increased ferritin content in Caco-2 cells at 25 μM for 2 hours
  • Showed a protective effect on colitis in mice at 500 mg/kg iron administered orally
Catalog No. 50-003-39933