Learn More
Medchemexpress LLC Ferrous bisglycinate | 20150-34-9 | 98% | 10 G
SDP
Supplier:  Medchemexpress LLC HY13007810G
Encompass_Preferred
Ferrous bisglycinate is an orally active iron fortificant and therapeutic iron supplement. It is primarily used for the research of iron deficiency anemia.
  • Functions as an orally active iron fortificant and therapeutic iron supplement.
  • Primarily used for the research of iron deficiency anemia.
  • In vitro studies show that it does not affect the viability of Caco-2 cells.
  • Increases ferritin content in Caco-2 cells.
  • Exerts a protective effect on colitis in mice.
  • Achieved 100% survival rates and caused the least body loss in an animal model of colitis.
Catalog No. 50-003-40965