Learn More
Waters Corp Waters Ferrule, 01, Stainless Steel, 10 pk
Supplier:  Waters Corp WAT005063
Encompass_Preferred
Tubing and fittings are meticulously designed to ensure leak-free connections with minimal dead volume and reduced band spreading. For optimal performance, these components must be clean and specifically engineered to work together. The geometry of the ferrule, the conical port it's used with, the tube diameter, and the fittings must all match. A variety of materials, lengths, and diameters are available to provide the necessary flexibility when setting up and maintaining a system.
  • Fitting type
  • Designed for HPLC systems
  • Made from stainless steel
  • Internal diameter of 1.5875 mm
  • Package contains 10 units
Catalog No. 50-819-008