Learn More
Microsolv Technology Corporation Ferrule, for use with Two Piece, High Pressure, Tubing Connectors for 1/16" PEEK Tubing. Universal Single Cone. 1 EA.
Supplier:  Microsolv Technology Corporation 4732036
Encompass_Preferred
This Single Cone Ferrule will swage on to the PEEK tubing in one place for a good grip on the tubing.Use with any PEEK, 2 Piece, 1/16" Tubing Connectors (Male Nuts).
Catalog No. 50-227-1504