Learn More
Chemglass Life Sciences Ferrule, Flangeless, 1/8", Yellow, Tefzel
SDP
Supplier:  Chemglass Life Sciences CG1164F02
Encompass_Preferred
Ferrule, Flangeless, 1/8", Yellow, Tefzel. Used with flangeless nut to ensure the tubing is held firmly in place. Ferrule designed to form a zero dead-volume connection. Ferrule is placed on the end of the tubing with the cone pointing toward the nut, flat end towards the PTFE fitting.
Catalog No. 50-872-343