Learn More
Trajan Scientific VICI Valco Ferrle, ZDV 10 Pack
Supplier:  Trajan Scientific BIO.ZF1S610
Encompass
VICI Valco Ferrule, Zero Dead Volue, ZDV, 1/16in, Stainless Steel, 10 Pack. Made in the USA.
Catalog No. NC0954190