Learn More
Medchemexpress LLC Ferulenol | 6805-34-1 | MFCD02183471 | 99.0% | 366.49 g·mol⁻¹ | C24H30O3 | 5 MG

Supplier: Medchemexpress LLC HY1296055MG
Ferulenol is a prenylated coumarin sesquiterpene derivative that specifically inhibits succinate ubiquinone reductase. It is supplied as a research reagent for in vitro and in vivo studies of mitochondrial metabolism and oxidative phosphorylation.
- High purity 99.0%.
- Chemical formula C24H30O3; molecular weight 366.49 g·mol⁻¹.
- Soluble ≥50 mg/mL in ethanol for in vitro use; suitable in vivo formulations reported.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Specifically inhibits succinate ubiquinone reductase, affecting mitochondrial oxidative phosphorylation.
- Available in small research sizes, such as 5 mg, 10 mg, and 50 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.