Learn More
Simmler Fetal Cell Stain Kit 300 Tests
Supplier:  Simmler S041200
Encompass_Preferred
Fetal Cell Stain Kit; Convenient flip-open spout caps; Provide complete staining in less than 20 min.; Includes three 120ml bottles of each reagent, Fixing Solution, Buffer Solution and the Fetal Cell Staining Solution; Exp: 18month; 300/PK
Catalog No. NC9822012