Learn More
Medchemexpress LLC Fetal-tau/0N3R Huma 5ug
SDP
Supplier:  Medchemexpress LLC HYP73618A5UG
Encompass_Preferred
Tau/0N3R protein is encoded by the Tau/0N3R gene and undergoes regulated alternative splicing to produce different mRNA species Its expression varies in the nervous system according to the maturation and type of neurons Fetal-tau/0N3R Protein Human (His) is the recombinant human-derived Fetal-tau/0N3R protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-79963