Learn More
QA Bio Inc Fetuin Glycoprotein Standard - 4 x 50µg
Supplier:  QA Bio Inc GCPFET50UX4
Encompass
The fetuin glycoprotein standard was developed for use during glycan release and labeling(particularly for DMB sialic acid analysis). Fetuin exists in a variety of glycoforms containing bi-, tri-, and tetra-antennary oligosaccharides with variable sialylation, as well as O-linked glycans. • Amount of Glycoprotein Supplied: GCP-FET-05 500 µg - GCP-FET-250U 250 µg - GCP-FET-50Ux4 50 µg per vial, 4 vials • Source: Fetal calf serum • Form Dry. Lyophilised powder. • Molecular Weight 36 kDa (protein weight only)
Catalog No. 50-223-6686