Learn More
TARGETMOL CHEMICALS INC FEVIPIPRANT 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T396525MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fevipiprant (NVP-QAW039) is a selective potent reversible competitive CRTh2 antagonist. purity: 99%
Catalog No. 50-258-0042