Learn More
TARGETMOL CHEMICALS INC Fexofenadine 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T21390100MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Fexofenadine (Carboxyterfenadine), an antihistamine pharmaceutical drug, is used to treat allergy symptoms, such as nasal congestion, hay fever, and urticaria. Compared to first-generation antihistamines, Fexofenadine is less able to pass the blood-brain barrier and cause sedation. Purity 99.87%
Catalog No. 50-287-7777