Learn More
Medchemexpress LLC Fexofenadine | 83799-24-0 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB080110MM/1ML
Encompass_Preferred
Fexofenadine is an orally active and nonsedative H1 receptor antagonist, suitable for research into allergic rhinitis and chronic idiopathic urticarial. It is provided as a 10 mM solution in DMSO.
  • Active and nonsedative H1 receptor antagonist
  • Can be used in allergic rhinitis research
  • Can be used in chronic idiopathic urticarial research
  • Purity of 99.8%
Catalog No. 50-004-08104