Learn More
Medchemexpress LLC Fexofenadine | 83799-24-0 | 99.8% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB08015MG
Encompass_Preferred
Fexofenadine is an orally active and nonsedative H1 receptor antagonist. It can be used in allergic rhinitis and chronic idiopathic urticarial research.
  • Orally active
  • Nonsedative H1 receptor antagonist
  • Used in allergic rhinitis research
  • Used in chronic idiopathic urticarial research
Catalog No. 50-004-09325