Learn More
Selleck Chemical LLC Fexofenadine HCl S3208-10mg
SDP
Supplier:  Selleck Chemical LLC S320810MG
Encompass_Preferred
Fexofenadine (MDL 16455A) inhibits histamine H1 receptor with IC50 of 246 nM
Catalog No. 50-310-9961