Learn More
Medchemexpress LLC Fexofenadine (hydrochloride) | 153439-40-8 | MFCD00865710 | 99.9% | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0801A10MM/1ML
Encompass_Preferred
Fexofenadine hydrochloride is an orally active and nonsedative H1 receptor antagonist. It is used in allergic rhinitis and chronic idiopathic urticarial research.
  • Orally active H1 receptor antagonist
  • Non-sedative
  • Used in allergic rhinitis research
  • Used in chronic idiopathic urticarial research
Catalog No. 50-004-09150