Learn More
TARGETMOL CHEMICALS INC Fexofenadine hydrochloride
SDP
Supplier:  TARGETMOL CHEMICALS INC T1470L50MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Fexofenadine Hydrochloride is a carboxylated metabolic derivative of terfenadine and long-lasting selective histamine H1 receptor antagonist, with antihistaminic activity. Purity 100%
Catalog No. 50-287-8263